Learn More
Grainger Radians Rad-Atac Safety Eyewear, Clear Polycarbonate, Wraparound, Anti-Fog, 1 ea.
Catalog No. 502785832
Encompass_Preferred
These wraparound safety glasses feature a lightweight design with a single polycarbonate lens and an integrated nosepiece for a secure and comfortable fit. Designed for reliable eye protection, the eyewear includes soft, non-slip rubber-tipped temples that enhance stability during extended wear, and they meet ANSI Z87.1+ and CSA Z94.3 high-impact standards.
  • Provides 99.9% UVA and UVB protection
  • Features anti-fog and hard-coat lens treatment
  • Constructed with a durable, impact-resistant polycarbonate lens
  • Offers a secure fit with an integrated nosepiece and soft rubber-tipped temples
  • Suitable for use in both indoor and outdoor environments
Catalog No. 50-278-5832 Supplier Grainger Supplier No. 199JC1
These wraparound safety glasses feature a lightweight design with a single polycarbonate lens and an integrated nosepiece for a secure and comfortable fit. Designed for reliable eye protection, the eyewear includes soft, non-slip rubber-tipped temples that enhance stability during extended wear, and they meet ANSI Z87.1+ and CSA Z94.3 high-impact standards.
  • Provides 99.9% UVA and UVB protection
  • Features anti-fog and hard-coat lens treatment
  • Constructed with a durable, impact-resistant polycarbonate lens
  • Offers a secure fit with an integrated nosepiece and soft rubber-tipped temples
  • Suitable for use in both indoor and outdoor environments