Learn More
MSC Radians Safety Eyewear, Anti-Fog, Polycarbonate, Clear, ANSI Z87.1, 1 ea
Catalog No. 5000684430
Encompass
The Mirage safety eyewear offers a lightweight, frameless, and wraparound design for comfortable and protective wear in various applications.
  • Sleek, lightweight design for a comfortable fit
  • Optically correct and distortion-free lenses
  • Economical dual wraparound 9.75 base lens provides near perfect orbital seal
  • Protective polycarbonate lens provides impact resistance
  • Weighs less than 1 oz.
  • Meets ANSI Z87.1 standards and provides 99.9% UV protection
  • Clear anti-fog lens for improved visibility
Catalog No. 50-006-84430 Supplier MSC Supplier No. 76169739
The Mirage safety eyewear offers a lightweight, frameless, and wraparound design for comfortable and protective wear in various applications.
  • Sleek, lightweight design for a comfortable fit
  • Optically correct and distortion-free lenses
  • Economical dual wraparound 9.75 base lens provides near perfect orbital seal
  • Protective polycarbonate lens provides impact resistance
  • Weighs less than 1 oz.
  • Meets ANSI Z87.1 standards and provides 99.9% UV protection
  • Clear anti-fog lens for improved visibility