Learn More
MSC Radians Safety Vest, Class 2 Breakaway, Large, 1 ea
Catalog No. 5000787205
Encompass
This safety vest is designed for high visibility and worker safety in environments with moving machinery or equipment. Its innovative breakaway design allows for quick removal to prevent entanglement and enhance safety.
  • ANSI/ISEA 107 Type R Class 2 rated
  • Five-point breakaway system at shoulders, sides, and front closure
  • Hook and loop fasteners for quick removal
  • Constructed from 100% polyester solid knit and mesh material
  • Two in silver reflective tape with one horizontal and two vertical stripes
Catalog No. 50-007-87205 Supplier MSC Supplier No. 69580199
This safety vest is designed for high visibility and worker safety in environments with moving machinery or equipment. Its innovative breakaway design allows for quick removal to prevent entanglement and enhance safety.
  • ANSI/ISEA 107 Type R Class 2 rated
  • Five-point breakaway system at shoulders, sides, and front closure
  • Hook and loop fasteners for quick removal
  • Constructed from 100% polyester solid knit and mesh material
  • Two in silver reflective tape with one horizontal and two vertical stripes