Learn More
MSC Radians Safety Vest, Class 2, Mesh, Zipper, X-Large, 1 ea
Catalog No. 5000685468
Encompass
The Radians SV2Z Economy Type R Class 2 Safety Vest with Zipper Closure offers compliant protection at an economical price. It is suitable for extreme work environments or short-term labor, providing durability and visibility.
  • Meets ANSI/ISEA 107 standards, Class 2
  • Zipper closure
  • Two in silver reflective tape
  • One horizontal stripe
  • One upper left front pocket
  • One lower right inside pocket
  • Constructed from 100% polyester solid knit and mesh materials
Catalog No. 50-006-85468 Supplier MSC Supplier No. 61523536
The Radians SV2Z Economy Type R Class 2 Safety Vest with Zipper Closure offers compliant protection at an economical price. It is suitable for extreme work environments or short-term labor, providing durability and visibility.
  • Meets ANSI/ISEA 107 standards, Class 2
  • Zipper closure
  • Two in silver reflective tape
  • One horizontal stripe
  • One upper left front pocket
  • One lower right inside pocket
  • Constructed from 100% polyester solid knit and mesh materials