Learn More
Alkali Scientific Radiant™ 2X GC-Long Mastermix 80 Reactions 2 x 1mL
SDP
Catalog No. 502389529
Encompass_Preferred
Radiant™ GC-Long Hot Start 2x Master Mix has been engineered for the amplification of extremely difficult templates and long amplicons up to35kb. Radiant™ GC-Long Hot Start 2x Master Mix possesses 5´-3´ DNA polymerase and 3´-5´ proofreading exonuclease activities with an errorrate of 4.55 x 10-7. The polymerase is ideal for cloning applications, library construction, screening and genotyping of problematic GC-rich andAT-rich sequences. Radiant™ GC-Long Hot Start 2x Master Mix Polymerase is engineered for robust, superior high-fidelity PCR in comparisonwith standard proof-reading enzymes such as pfu DNA Polymerase. Several point-mutations combined with a new-generation bufferformulation provide for significantly improved processivity for faster cycling, greater sensitivity and less inhibition with crude DNA samples.
Catalog No. 50-238-9529 Supplier Alkali Scientific Supplier No. C550
Radiant™ GC-Long Hot Start 2x Master Mix has been engineered for the amplification of extremely difficult templates and long amplicons up to35kb. Radiant™ GC-Long Hot Start 2x Master Mix possesses 5´-3´ DNA polymerase and 3´-5´ proofreading exonuclease activities with an errorrate of 4.55 x 10-7. The polymerase is ideal for cloning applications, library construction, screening and genotyping of problematic GC-rich andAT-rich sequences. Radiant™ GC-Long Hot Start 2x Master Mix Polymerase is engineered for robust, superior high-fidelity PCR in comparisonwith standard proof-reading enzymes such as pfu DNA Polymerase. Several point-mutations combined with a new-generation bufferformulation provide for significantly improved processivity for faster cycling, greater sensitivity and less inhibition with crude DNA samples.