Learn More
Alkali Scientific Radiant™ cDNA Synthesis Kit, 1-Step, 5 Reactions, 1 x 0.2mL 5X cDNA Mix, 1 x 0.005mL 20X RTase
SDP
Catalog No. 503029031
Encompass_Preferred
Radiant™ cDNA Synthesis Kit, 1-Step • Unbiased representation of 5’ and 3’ mRNA transcript ends• Sensitive detection of low copy number transcripts• High cDNA yields from as little as 4pg total RNA• Simple 2 tube systemSUPPLIED WITH a proprietary ratio of anchored oligo dT and random hexamers
Catalog No. 50-302-9031 Supplier Alkali Scientific Supplier No. QC1125S
Radiant™ cDNA Synthesis Kit, 1-Step • Unbiased representation of 5’ and 3’ mRNA transcript ends• Sensitive detection of low copy number transcripts• High cDNA yields from as little as 4pg total RNA• Simple 2 tube systemSUPPLIED WITH a proprietary ratio of anchored oligo dT and random hexamers