Learn More
Alkali Scientific Radiant DNA Extract Amplify T
SDP
Catalog No. 503026526
Encompass_Preferred
Radiant DNA Extract and Amplify Tissue PCR kit combines rapid DNA extraction with fast highly specific DNA amplification in a convenient easy to use format Eliminate the need for overnight and time intensive DNA extraction methods with this simple integrated extraction and amplification PCR kit powered by the latest advances in hot-start polymerase technology Radiant Extract and Amplify Tissue PCR Kit has been developed for fast efficient amplification of DNA from a variety of tissues and is particularly suited to solid tissue such as mouse tail or mouse ear DNA extraction is performed in a single tube removing the need for multiple washing steps Extraction of DNA is rapid providing DNA for PCR in 15 minutes Extraction takes place in a single tube minimizing potential contamination Extracted DNA is amplified using supplied 2X Radiant RED Hot-Start Taq Mix Our antibody hot start polymerase uses the latest developments in polymerase technology and buffer chemistry to enhance PCR
Catalog No. 50-302-6526 Supplier Alkali Scientific Supplier No. C462S
Radiant DNA Extract and Amplify Tissue PCR kit combines rapid DNA extraction with fast highly specific DNA amplification in a convenient easy to use format Eliminate the need for overnight and time intensive DNA extraction methods with this simple integrated extraction and amplification PCR kit powered by the latest advances in hot-start polymerase technology Radiant Extract and Amplify Tissue PCR Kit has been developed for fast efficient amplification of DNA from a variety of tissues and is particularly suited to solid tissue such as mouse tail or mouse ear DNA extraction is performed in a single tube removing the need for multiple washing steps Extraction of DNA is rapid providing DNA for PCR in 15 minutes Extraction takes place in a single tube minimizing potential contamination Extracted DNA is amplified using supplied 2X Radiant RED Hot-Start Taq Mix Our antibody hot start polymerase uses the latest developments in polymerase technology and buffer chemistry to enhance PCR