Learn More
Alkali Scientific Radiant DNA Markers and Ladder
SDP
Supplier:  Alkali Scientific C617
Encompass_Preferred
A ready-to-use molecular grade dNTP Mix containing dATP dCTP dGTP and dTTP at pH 7 5 as sodium salts in purified water The DNA Markers and Ladders I-IV is designed to save hands-on time for researchers and minimize the possibility of contamination For use in DNA polymerization reactions DNA labeling and sequencing processes Reliable PCR grade All ASI dNTPs are supplied as sodium salts in purified water at pH 7 5 Sodium salts have resistance to repeated freezing and thawing cycles Also dNTP preparations remain sterile over the entire shelf-life due to the bacteriostatic activity of sodium towards various microorganisms dNTPs Mol Wt Molar Extinction(1 mol) dATP 514 9 15 4 (A259) dGTP 530 9 13 7 (A252) dCTP 490 9 9 1 (A272) dTTP 505 9 9 6 (A267) Convenient pre-optimized and pre-mixed Ultra-pure 99% trisphosphate by HPLC Extended shelf-life of 18 months at -20C Free from PCR inhibitors DNase RNase and Nickase free Manufactured by ASI in a purpose-built facility Applications St
Catalog No. 50-302-6421