Learn More
Alkali Scientific Radiant™ HiFi Polymerase Pro™ 2X RED Master Mix, w/ Buffer, dNTPs, & PCR enhancers, 1 x 1.25 mL (50 Reactions)
SDP
Catalog No. 503027030
Encompass_Preferred
Radiant™ HiFi Polymerase Pro™ 2X RED Mastermix is a versatile and robust high fidelity DNA polymerase engineered for all PCR applications where greater sequence accuracy is required. Improved DNA binding and increased processivity result in shorter extension times, higher yields and the ability to amplify longer and more difficult targets. High temperature cycling – up to 100 °C denaturation to better separate GC-rich templates Increased PCR success rates with complex genomic templates (17.5 kb and over) High yields under standard and fast PCR conditions (10-30 s/kb) Efficient and specific amplification from challenging templates including GC and AT-rich sequences 100x higher fidelity than Taq DNA polymerase Generates blunt-end PCR products Available as a convenient 2x ready mix with the option of a red dye for direct gel loading
Catalog No. 50-302-7030 Supplier Alkali Scientific Supplier No. HFP810
Radiant™ HiFi Polymerase Pro™ 2X RED Mastermix is a versatile and robust high fidelity DNA polymerase engineered for all PCR applications where greater sequence accuracy is required. Improved DNA binding and increased processivity result in shorter extension times, higher yields and the ability to amplify longer and more difficult targets. High temperature cycling – up to 100 °C denaturation to better separate GC-rich templates Increased PCR success rates with complex genomic templates (17.5 kb and over) High yields under standard and fast PCR conditions (10-30 s/kb) Efficient and specific amplification from challenging templates including GC and AT-rich sequences 100x higher fidelity than Taq DNA polymerase Generates blunt-end PCR products Available as a convenient 2x ready mix with the option of a red dye for direct gel loading