Learn More
Alkali Scientific Radiant™ Hot Start Taq DNA Polymerase for FAM & SYBR green dye users constructing qPCR mix, 250 Reactions, (1 x .05mL Hot-Start Taq Polymerase, 1 x 1.5mL 20X KCI Reaction Buffer)
SDP
Catalog No. 503026624
Encompass_Preferred
Radiant™ Hot Start Taq DNA Polymerase for FAM & SYBR green dye users constructing qPCR mix Radiant™ Hot-Start Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to enhance PCR speed, yield and specificity. Radiant™ Hot-Start Taq DNA Polymerase is especially suited to application where absence of residual host bacterial DNA is critical. Our HS Taq has an error rate of approximately 1 error per 2.0 x 105 nucleotides incorporated. PCR products generated with Radiant™ Hot-Start Taq Polymerase are A-tailed and may be cloned into TA cloning vectors.
Catalog No. 50-302-6624 Supplier Alkali Scientific Supplier No. C321K
Radiant™ Hot Start Taq DNA Polymerase for FAM & SYBR green dye users constructing qPCR mix Radiant™ Hot-Start Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to enhance PCR speed, yield and specificity. Radiant™ Hot-Start Taq DNA Polymerase is especially suited to application where absence of residual host bacterial DNA is critical. Our HS Taq has an error rate of approximately 1 error per 2.0 x 105 nucleotides incorporated. PCR products generated with Radiant™ Hot-Start Taq Polymerase are A-tailed and may be cloned into TA cloning vectors.