Learn More
Alkali Scientific Radiant™ HRM Mix, High resolution melt - SNP detection, 100 Reactions, (1 x 1mL)
SDP
Catalog No. 503029287
Encompass_Preferred
Radiant™HRM Mix produces ultra-sensitive melt profiles allowing accurate discrimination subtle sequence variations including class I to class IV mutations and CpG methylation differences. Fast and accurate detection of subtle sequence differences Ultra-sensitive fluorescence profiles for clear characterisation of samples Third generation saturating dye SyGreen 2 Antibody-mediated hot start technology for improved sensitivity Compatible with all real-time instruments with HRM capability
Catalog No. 50-302-9287 Supplier Alkali Scientific Supplier No. QHRM1
Radiant™HRM Mix produces ultra-sensitive melt profiles allowing accurate discrimination subtle sequence variations including class I to class IV mutations and CpG methylation differences. Fast and accurate detection of subtle sequence differences Ultra-sensitive fluorescence profiles for clear characterisation of samples Third generation saturating dye SyGreen 2 Antibody-mediated hot start technology for improved sensitivity Compatible with all real-time instruments with HRM capability