Learn More
Alkali Scientific Radiant™ Library Quant Kit for Illumina Separate ROX, w/ Standards, Primers & Buffer, 100 x 20μL Reactions
SDP
Catalog No. 5000662708
Encompass_Preferred
Radiant™ Library Quant Kit for Illumina Separate ROX, with Standards, Primers & Buffer. Includes 100 x 20μL Reactions. This kit is optimized for Illumina platforms using Separate ROX, providing highly accurate and reproducible results for next-generation sequencing. The carefully balanced combination of primers, standards, and buffer allows for precise library quantification, making it suitable for a wide range of genomic and sequencing applications.
Catalog No. 50-006-62708 Supplier Alkali Scientific Supplier No. NGS0001
Radiant™ Library Quant Kit for Illumina Separate ROX, with Standards, Primers & Buffer. Includes 100 x 20μL Reactions. This kit is optimized for Illumina platforms using Separate ROX, providing highly accurate and reproducible results for next-generation sequencing. The carefully balanced combination of primers, standards, and buffer allows for precise library quantification, making it suitable for a wide range of genomic and sequencing applications.