Learn More
Alkali Scientific Radiant™ Probe 1-Step Lo-Rox qPCR Virus Kit, 1000 x 20µl Reactions, 4X
SDP
Catalog No. 503029036
Encompass_Preferred
Recommended kit for SARS-CoV-2 diagnostics Validated for qualitative detection of SARS-CoV-2 nucleic acid Concentrated 4x mix format, ideal for high-throughput, highly multiplexed assays Ultra-sensitive detection of RNA viruses Earlier detection of a wide range of template amounts Includes UltraScript RTase for thermostable reverse transcription up to 55°C Advanced RNase inhibitor Antibody-mediated hot start technology Compatible with all real-time PCR platforms – standard and fast cycling conditions The kit has been optimised with a high-concentration 4x mix, enabling greater sample input and increasing sensitivity, even when working with small volume reactions.
Catalog No. 50-302-9036 Supplier Alkali Scientific Supplier No. QL4098
Recommended kit for SARS-CoV-2 diagnostics Validated for qualitative detection of SARS-CoV-2 nucleic acid Concentrated 4x mix format, ideal for high-throughput, highly multiplexed assays Ultra-sensitive detection of RNA viruses Earlier detection of a wide range of template amounts Includes UltraScript RTase for thermostable reverse transcription up to 55°C Advanced RNase inhibitor Antibody-mediated hot start technology Compatible with all real-time PCR platforms – standard and fast cycling conditions The kit has been optimised with a high-concentration 4x mix, enabling greater sample input and increasing sensitivity, even when working with small volume reactions.