Learn More
Alkali Scientific Radiant™ Probe 1-Step No-ROX qPCR Kit, 1200 Reactions, (12 X 1mL 2X Mix, 12 X 0.2mL 20X RTase)
SDP
Catalog No. 502388721
Encompass_Preferred
Alkali Scientific offers Radiant™ Probe 1-Step No-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step No-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.
Catalog No. 50-238-8721 Supplier Alkali Scientific Supplier No. QN6012
Alkali Scientific offers Radiant™ Probe 1-Step No-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step No-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.