Learn More
Alkali Scientific Radiant™ Probe 1Step LoRox qPCR Virus Kit, 4X
SDP
Catalog No. 502388886
Encompass_Preferred
cDNA Synthesis and qPCR in a single tubeHighly concentrated 4X mix allows for increased sample and viral input. This optimizes sample sensitivity with small reaction volumes.Engineered for use with a wide range of probe technologies including TaqMan, Scorpions and molecular beacon probesMix includes the thermostable Radiant™ Reverse Transcriptase which is blended with an advanced RNase inhibitor to prevent degradation of RNA by contaminating RNasePlease see product attachment(s)
Catalog No. 50-238-8886 Supplier Alkali Scientific Supplier No. QL4099
cDNA Synthesis and qPCR in a single tubeHighly concentrated 4X mix allows for increased sample and viral input. This optimizes sample sensitivity with small reaction volumes.Engineered for use with a wide range of probe technologies including TaqMan, Scorpions and molecular beacon probesMix includes the thermostable Radiant™ Reverse Transcriptase which is blended with an advanced RNase inhibitor to prevent degradation of RNA by contaminating RNasePlease see product attachment(s)