Learn More
Alkali Scientific Radiant™ Probe qPCR Kit, 5,000 Reactions, (50 x 1mL), LoROX
SDP
Catalog No. 503029018
Encompass_Preferred
Radiant™ Probe qPCR Kit Novel hot-start chemistry for improved specificity and sensitivity. Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed. Broad range detection for increased reliability in low-copy assays Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods
Catalog No. 50-302-9018 Supplier Alkali Scientific Supplier No. QP9050
Radiant™ Probe qPCR Kit Novel hot-start chemistry for improved specificity and sensitivity. Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed. Broad range detection for increased reliability in low-copy assays Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods