Learn More
Alkali Scientific Radiant™ Probe qPCR Mix, Separate ROX, 500 Reactions, 5 x 1.0mL
SDP
Catalog No. 5000662954
Encompass_Preferred
Radiant™ Probe qPCR Mix, Separate ROX, 500 Reactions, 5 x 1.0mL, is a high-efficiency mix designed for real-time PCR analysis. The separate ROX feature ensures reliable signal normalization, while the 2x formulation provides optimal amplification efficiency. This product is ideal for high-throughput gene expression analysis, offering both sensitivity and accuracy. Whether you're studying gene expression or detection, this mix is an excellent choice for a range of PCR applications.
Catalog No. 50-006-62954 Supplier Alkali Scientific Supplier No. QP0005
Radiant™ Probe qPCR Mix, Separate ROX, 500 Reactions, 5 x 1.0mL, is a high-efficiency mix designed for real-time PCR analysis. The separate ROX feature ensures reliable signal normalization, while the 2x formulation provides optimal amplification efficiency. This product is ideal for high-throughput gene expression analysis, offering both sensitivity and accuracy. Whether you're studying gene expression or detection, this mix is an excellent choice for a range of PCR applications.