Learn More
Alkali Scientific Radiant™ qPCR Genotyping Mixgenotyping mix for qPCR, Hi-Rox, 100 Reactions, (1 X 1mL)
SDP
Catalog No. 502388678
Encompass_Preferred
Using the latest advancements in polymerase technology and advanced buffer chemistry Radiant™ Genotyping Mix Hi-Rox offers market leading performance with minimal optimization. Radiant™ Genotyping Mix Hi-Rox is a kit designed for use in dual-labeled probe based genotyping assays including Taqman, Molecular Beacon and Scorpion probe genotyping. Radiant™ Genotyping Mix is fully compatible with ABI TaqMan Pre-Designed SNP Genotyping assays. Radiant™ Genotyping Mix uses proprietary smallmolecular inhibitor technology that prevents formation of primer-dimers to improve reaction sensitivity and specificity. Highthroughput screening has resulted in a buffer system that allows efficient amplification from GC-rich and AT-rich templates, under fast and standard cycling conditions.
Catalog No. 50-238-8678 Supplier Alkali Scientific Supplier No. QG2001
Using the latest advancements in polymerase technology and advanced buffer chemistry Radiant™ Genotyping Mix Hi-Rox offers market leading performance with minimal optimization. Radiant™ Genotyping Mix Hi-Rox is a kit designed for use in dual-labeled probe based genotyping assays including Taqman, Molecular Beacon and Scorpion probe genotyping. Radiant™ Genotyping Mix is fully compatible with ABI TaqMan Pre-Designed SNP Genotyping assays. Radiant™ Genotyping Mix uses proprietary smallmolecular inhibitor technology that prevents formation of primer-dimers to improve reaction sensitivity and specificity. Highthroughput screening has resulted in a buffer system that allows efficient amplification from GC-rich and AT-rich templates, under fast and standard cycling conditions.