Learn More
Alkali Scientific Radiant™ Reverse Transcriptase, 40,000 Units
SDP
Catalog No. 503028955
Encompass_Preferred
Radiant™ Reverse Transcriptase is a high-performance enzyme designed for efficient and accurate cDNA synthesis from RNA templates. Engineered for superior sensitivity and reliability, it enables optimal reverse transcription for applications like RT-qPCR, RNA sequencing, and gene expression analysis. Ideal for low-input or degraded RNA samples, Radiant™ ensures robust performance with minimal background, delivering consistent and reproducible results. Unbiased representation of 5’ and 3’ mRNA transcript ends Reduced RNase H activity  Thermostable reverse transcriptase 45 °C to 55 °C Advanced RNase inhibitor High cDNA yields from as little as 4 pg total RNA Accurate reverse transcription of GC-rich templates Sensitive detection of low copy number transcripts Advanced buffer chemistry including Mg and dNTPs
Catalog No. 50-302-8955 Supplier Alkali Scientific Supplier No. QRT04
Radiant™ Reverse Transcriptase is a high-performance enzyme designed for efficient and accurate cDNA synthesis from RNA templates. Engineered for superior sensitivity and reliability, it enables optimal reverse transcription for applications like RT-qPCR, RNA sequencing, and gene expression analysis. Ideal for low-input or degraded RNA samples, Radiant™ ensures robust performance with minimal background, delivering consistent and reproducible results. Unbiased representation of 5’ and 3’ mRNA transcript ends Reduced RNase H activity  Thermostable reverse transcriptase 45 °C to 55 °C Advanced RNase inhibitor High cDNA yields from as little as 4 pg total RNA Accurate reverse transcription of GC-rich templates Sensitive detection of low copy number transcripts Advanced buffer chemistry including Mg and dNTPs