Learn More
Alkali Scientific Radiant™ SYBR Green Fluorescein qPCR Kit, 500 Reactions, (5 x 1mL)
SDP
Catalog No. 503028306
Encompass_Preferred
Radiant™ SYBR Green Flourescein qPCR Kit: Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed. Novel hot-start chemistry for improved specificity and sensitivity. Broad range detection for increased reliability in low-copy assays Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods Compatible with BioRad iCycler™, MyIQ™ & IQ5™  
Catalog No. 50-302-8306 Supplier Alkali Scientific Supplier No. QS3005
Radiant™ SYBR Green Flourescein qPCR Kit: Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed. Novel hot-start chemistry for improved specificity and sensitivity. Broad range detection for increased reliability in low-copy assays Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods Compatible with BioRad iCycler™, MyIQ™ & IQ5™  