Learn More
Alkali Scientific Radiant™ SYBR Green Master Mix qPCR Kit, 5000 Reactions, 50 x 1mL, HiROX
SDP
Catalog No. 503028956
Encompass_Preferred
Radiant™ SYBR Green Master Mix qPCR Kit • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed.• Novel hot-start chemistry for improved specificity and sensitivity.• Broad range detection for increased reliability in low-copy assays• Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods
Catalog No. 50-302-8956 Supplier Alkali Scientific Supplier No. QS2050
Radiant™ SYBR Green Master Mix qPCR Kit • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed.• Novel hot-start chemistry for improved specificity and sensitivity.• Broad range detection for increased reliability in low-copy assays• Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods