Learn More
Alkali Scientific Radiant™ 2X RED Hot-Start Taq Mix 200 Reactions 5 x 1mL
SDP
Supplier:  Alkali Scientific C441
Encompass_Preferred
RadiantTM Red 2X Hot-Start Taq Mastermix is a ready-to-use, robust PCR mix containing our highly purified Hot-Start Taq DNA Polymerase, reaction buffer, ultra-pure dNTPs, MgCl2 and PCR enhancers. RadiantTM Red 2X Hot Start Taq Mastermix is optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian & any crude genomic DNA and is ideal for screening, colony PCR, high-throughput PCR and genotyping of problematic GC-rich and AT-rich sequences. The Hot-Start technology is based on a new-generation PCR buffer formulation in conjunction with a proprietary antibody-mediated chemistry. RadiantTM Red 2X Taq Hot-Start Mastermix is provided at 2× concentration and used at 1× concentration by adding template, primer, and H2O. • Intert red dye will not inhibit PCR reaction & is used as a downstream Gel Loading Dye when running gel • No change in PCR cycling conditions when in supplied Red format. Red Dye cycles within PCR machine • New-gener
Catalog No. 50-233-5810