Learn More
Alkali Scientific Radiant™ 2X RED Taq Mastermix, 1000 Reactions, (25 X 1mL)
SDP
Catalog No. 502335997
Encompass_Preferred
Radiant™ Red 2X Taq Mastermix is optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA and is ideal for screening, colony PCR, high-throughput PCR and genotyping of problematic GC-rich and AT-rich sequences. Radiant™ª Red 2X Taq Mastermix is provided at 2× concentration and used at 1× concentration by adding template, primer, and H2O.
Catalog No. 50-233-5997 Supplier Alkali Scientific Supplier No. C225
Radiant™ Red 2X Taq Mastermix is optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA and is ideal for screening, colony PCR, high-throughput PCR and genotyping of problematic GC-rich and AT-rich sequences. Radiant™ª Red 2X Taq Mastermix is provided at 2× concentration and used at 1× concentration by adding template, primer, and H2O.