Learn More
Alkali Scientific Radiant™ cDNA Synthesis Kit, 1-Step, 100 Reactions, (4 x 0.1mL 5X cDNA Mix, 4 x 0.025mL 20X RTase)
SDP
Catalog No. 502335767
Encompass_Preferred
The Radiant™ cDNA Synthesis Kit uses the latest developments in reverse transcriptase technology and buffer chemistry to enhance cDNA synthesis speed and yield with accurate transcript representation. The reverse transcriptase, buffer system and combination of random hexamers with anchored oligo(dT) allow for unbiased, efficient, sensitive cDNA synthesis.High quality cDNA synthesis for downstream qPCR analysis is essential for successful expression studies. Many factors affect cDNA synthesis including the reverse transcriptase, buffer systems, enhancers and priming strategy. The Radiant™ cDNA synthesis mix removes the need for user optimization of these critical factors.
Catalog No. 50-233-5767 Supplier Alkali Scientific Supplier No. QC1155
The Radiant™ cDNA Synthesis Kit uses the latest developments in reverse transcriptase technology and buffer chemistry to enhance cDNA synthesis speed and yield with accurate transcript representation. The reverse transcriptase, buffer system and combination of random hexamers with anchored oligo(dT) allow for unbiased, efficient, sensitive cDNA synthesis.High quality cDNA synthesis for downstream qPCR analysis is essential for successful expression studies. Many factors affect cDNA synthesis including the reverse transcriptase, buffer systems, enhancers and priming strategy. The Radiant™ cDNA synthesis mix removes the need for user optimization of these critical factors.