Learn More
Alkali Scientific Radiant™ Extract & Amplify Tissue PCR Kit 400 Reactions 10 x 1mL 2X HS Taq Red, 5 x 1.6mL 5X Extract Buffer A, 5 x .08mL 10X Extract Buffer B
SDP
Catalog No. 502335916
Encompass_Preferred
Radiant™ Extract and Amplify Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tail, ear punch and other mammalian tissues, buccal swabs, hair shafts, and saliva. SPEED UP your GENOTYPING NO OVERNIGHT lysing Protocol: Extraction Solutions, included: 15 minutes then PCR as directed with supplied Radiant™ RED 2X mix During the quick 5 minute extraction process, DNA is released from the starting material by mixing the sample with ASI's proprietary Extraction Solution. This is followed by a 10 minute incubation at 95 degree to help inactivate proteins and genomic DNA. NO OVERNIGHT EXTRACTION. There is no need for mechanical disruption, organic extraction, column purification, or precipitation of the DNA.
Catalog No. 50-233-5916 Supplier Alkali Scientific Supplier No. C465
Radiant™ Extract and Amplify Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tail, ear punch and other mammalian tissues, buccal swabs, hair shafts, and saliva. SPEED UP your GENOTYPING NO OVERNIGHT lysing Protocol: Extraction Solutions, included: 15 minutes then PCR as directed with supplied Radiant™ RED 2X mix During the quick 5 minute extraction process, DNA is released from the starting material by mixing the sample with ASI's proprietary Extraction Solution. This is followed by a 10 minute incubation at 95 degree to help inactivate proteins and genomic DNA. NO OVERNIGHT EXTRACTION. There is no need for mechanical disruption, organic extraction, column purification, or precipitation of the DNA.