Learn More
Alkali Scientific Radiant™ Green Hi-ROX qPCR Kit, 100 Reactions, (1 x 1mL)
SDP
Catalog No. 502335766
Encompass_Preferred
Alkali Scientific offers Radiant™ Green Hi-ROX qPCR Kits, a high-performance 2X ready-to-use real-time PCR reagents designed for best-in-class quantitative PCR. Radiant™ Green Hi-ROX qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Green Hi-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection in combination with fluorescein for optional use with certain qPCR instruments.
Catalog No. 50-233-5766 Supplier Alkali Scientific Supplier No. QS2001
Alkali Scientific offers Radiant™ Green Hi-ROX qPCR Kits, a high-performance 2X ready-to-use real-time PCR reagents designed for best-in-class quantitative PCR. Radiant™ Green Hi-ROX qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Green Hi-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection in combination with fluorescein for optional use with certain qPCR instruments.