Learn More
Alkali Scientific Radiant™ Probe Lo-ROX qPCR Kit, 2,000 Reactions, (20 x 1mL)
SDP
Catalog No. 502335930
Encompass_Preferred
Alkali Scientific offers Radiant™ Probe Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in-class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. Furthermore, Radiant™ Probe Lo-ROX qPCR Kits are optimized for advanced multiplexing capability, delivering the same high efficiency, early Ct, and reaction speed even in complex multiplex assays.
Catalog No. 50-233-5930 Supplier Alkali Scientific Supplier No. QP9020
Alkali Scientific offers Radiant™ Probe Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in-class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. Furthermore, Radiant™ Probe Lo-ROX qPCR Kits are optimized for advanced multiplexing capability, delivering the same high efficiency, early Ct, and reaction speed even in complex multiplex assays.