Learn More
Alkali Scientific Radiant™ Taq DNA Polymerase, 5u/µl, 500 units w/ separate tube 10X PCR buffer and MgCl2
SDP
Catalog No. 502335989
Encompass_Preferred
Radiant™ Taq DNA Polymerase Description RadiantTM Taq DNA Polymerase is a highly purified, high performance DNA Polymerase optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA. RadiantTM Taq DNA Polymerase exhibits 5´-3´ DNA polymerase activity with an error-rate of wild-type Taq ( 2.0 x 10-5). The polymerase is ideal for screening, colony PCR, high-throughput PCR and genotyping of problematic GC-rich and AT-rich sequences. RadiantTM Taq DNA Polymerase is engineered for robust, superior PCR and is supplied with a highly optimized, new-generation buffer system which provides exceptional sensitivity. Storage RadiantTM Taq DNA Polymerase is shipped on blue or dry ice and should be stored at –20°C upon receipt. Excessive freeze/thawing should be avoided. When stored as specified, RadiantTM Taq DNA Polymerase is stable for 12 months from date of receipt. The Kit may also be stored at 4°C for 1 month. Important Cons
Catalog No. 50-233-5989 Supplier Alkali Scientific Supplier No. C101
Radiant™ Taq DNA Polymerase Description RadiantTM Taq DNA Polymerase is a highly purified, high performance DNA Polymerase optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA. RadiantTM Taq DNA Polymerase exhibits 5´-3´ DNA polymerase activity with an error-rate of wild-type Taq ( 2.0 x 10-5). The polymerase is ideal for screening, colony PCR, high-throughput PCR and genotyping of problematic GC-rich and AT-rich sequences. RadiantTM Taq DNA Polymerase is engineered for robust, superior PCR and is supplied with a highly optimized, new-generation buffer system which provides exceptional sensitivity. Storage RadiantTM Taq DNA Polymerase is shipped on blue or dry ice and should be stored at –20°C upon receipt. Excessive freeze/thawing should be avoided. When stored as specified, RadiantTM Taq DNA Polymerase is stable for 12 months from date of receipt. The Kit may also be stored at 4°C for 1 month. Important Cons