Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shamrock Labels Radiation Warning Labels
SDP

Catalog No. 1187311 Shop All Shamrock Scientific Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-873-11 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-873-11 Supplier Shamrock Labels Supplier No. SRA11
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used on any surface where visual warning is required

  • Ideal for labeling tubes, flasks, pipetters and tips
  • 1/4 in. wide (0.6cm) label printed with vivid magenta ink on bright yellow background
  • Glove-friendly permanent adhesive holds labels firmly in place
  • Roll fits standard label dispensers

Specifications

Adhesive Type Permanent
Color Yellow
Format Label
Material Litho
Product Type Label
For Use With (Application) Ideal for labeling tubes, flasks, pipetters and tips
Quantity 1000/Roll
Width (English) 0.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.