Learn More
Research Products International Corp Radicicol [Humicola Fuscoatra], 1 Milligram
SDP
Catalog No. 50488976
Encompass_Preferred
Radicicol is a naturally occurring antibiotic and antifungal compound . It is known for its ability to inhibit the activity of heat shock protein 90 (Hsp90), a molecular chaperone that plays a crucial role in the folding and stabilization of various client proteins involved in cell signaling and regulation. 
Radicicol is often used as a tool to study Hsp90-dependent processes and the functions of client proteins. Researchers use radicicol to investigate the impact of inhibiting Hsp90 on cellular pathways, protein folding, and cell viability. This compound is particularly valuable in cancer research, as many oncogenic proteins are dependent on Hsp90 for proper folding and stability.
Catalog No. 50-488-976 Supplier Research Products International Corp Supplier No. R200200.001
Radicicol is a naturally occurring antibiotic and antifungal compound . It is known for its ability to inhibit the activity of heat shock protein 90 (Hsp90), a molecular chaperone that plays a crucial role in the folding and stabilization of various client proteins involved in cell signaling and regulation. 
Radicicol is often used as a tool to study Hsp90-dependent processes and the functions of client proteins. Researchers use radicicol to investigate the impact of inhibiting Hsp90 on cellular pathways, protein folding, and cell viability. This compound is particularly valuable in cancer research, as many oncogenic proteins are dependent on Hsp90 for proper folding and stability.