Learn More
Medchemexpress LLC HY-N6769 1mg Medchemexpress, Radicicol CAS:12772-57-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67691MG
Encompass_Preferred
Medchemexpress, HY-N6769 1mg Radicicol CAS:12772-57-5 Radicicol is an inhibitor of Hsp90 with an IC50 value of 1 μM. Radicicol binds to the ATPase domain of Hsp90 and prevents maturation of Hsp90 clients, leading to proteasomal degradation. Radicicol is an antifungal antibiotic with antimalarial activity, impairs mitochondrial replication by targeting P. falciparum topoisomerase VIB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0465