Learn More
Abcam Radicicol, Heat shock protein (HSP) 90 inhibitor, 1MG
Catalog No. 502706410
Encompass_Preferred
MW 364.8 Da, Purity >99%. Heat shock protein 90 inhibitor and antifungal antibiotic. Binds to the N-terminal ATP/ADP-binding domain suppressing cell transformation induced by src, ras and mos. Induces cell cycle arrest at G₁-S phase. Shows antiangiogenic effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6410 Supplier Abcam Supplier No. AB1418481MG
MW 364.8 Da, Purity >99%. Heat shock protein 90 inhibitor and antifungal antibiotic. Binds to the N-terminal ATP/ADP-binding domain suppressing cell transformation induced by src, ras and mos. Induces cell cycle arrest at G₁-S phase. Shows antiangiogenic effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement