Learn More
MilliporeSigma™ Supelco™ radiello™ Barcode Labels
Catalog No. 111011419 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
198/Pk.
Catalog No. Quantity
11-101-1419 198/Pk.
1 options
Catalog No. 11-101-1419 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD190
Add to Cart
Add to Cart
Each self-adhesive label allows users to record sampling date/time and displays a unique barcode for the unmistakable identification of the cartridge adsorbent during sampling, desorption, and analysis.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Air Monitoring
Quantity 198/Pk.
radiello is a trademark of Institi Clinici Scientifici Maugeri