Learn More
MilliporeSigma™ Supelco™ radiello™ Diffusive Bodies
Catalog No. 111008562 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Anaesthetic Gases and Vapors
General Use
Reduced Sampling Rates
Sampling Light Sensitive Compounds
Material:
Polyethylene
Silicone
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Material
11-100-8562 Anaesthetic Gases and Vapors Silicone
11-100-9412 Sampling Light Sensitive Compounds Polyethylene
11-100-9514 General Use Polyethylene
11-100-9056 Reduced Sampling Rates Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-100-8562 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD1203
Only null left
Add to Cart
Add to Cart
Designed to house radiello™ cartridge adsorbents during sampling
Diffusive bodies are threaded at one end for easy attachment to the radiello™ triangular support plate. Unlike most of the cartridge adsorbents, the diffusive bodies are reusable and can be cleaned with a mild detergent between use. The diffusive bodies will collect dust (especially during outdoor sampling) and replacement is recommended after 4-5 uses. Dimensions are 60 mm height x 16 mm diameter. There are four different diffusive bodies available. Each of which are application specific in design.
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone
For Use With (Application) Anaesthetic Gases and Vapors
Quantity 20/Pk.
radiello is a trademark of Institi Clinici Scientifici Maugeri