Learn More
MilliporeSigma™ Supelco™ radiello™ Filtration Kit
Catalog No. 111010482 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
11-101-0482 20/Pk.
1 options
Catalog No. 11-101-0482 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD174
Add to Cart
Add to Cart
Ideal for filtering aqueous samples prior to reversed-phase HPLC and ion-chromatography
The filtration kit consists of graduated polypropylene syringe barrels and 13 mm diameter syringe filters with 0.45 μm porosity. The filtration kit is ideal for filtering aqueous samples prior to reversed-phase HPLC and ion-chromatography.
TRUSTED_SUSTAINABILITY
Specifications
Includes Graduated polypropylene syringe barrels, 13 mm diameter syringe filters with 0.45 μm Pore Size
Quantity 20/Pk.
Type Filtration Kit
radiello is a trademark of Institi Clinici Scientifici Maugeri