Learn More
MilliporeSigma™ Supelco™ radiello™ Hydrogen Sulfide Starter Kit
Catalog No. 111010793 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
11-101-0793 1 Kit
1 options
Catalog No. 11-101-0793 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD170S
Only null left
Add to Cart
Add to Cart
radiello™ Starter Kits are ideal for introducing you to the product and include everything you need to take one complete sample. No additional parts are required.
Kit includes: 2 RAD170 adsorbing cartridges and barcode labels, 1 RAD120 white diffusive body, 1 RAD121 triangular support plate and 1 instruction sheet
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Air Monitoring
Includes 2 RAD170 adsorbing cartridges and barcode labels, 1 RAD120 white diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet
Quantity 1 Kit
radiello is a trademark of Institi Clinici Scientifici Maugeri