Learn More
MilliporeSigma™ Supelco™ radiello™ Outdoor Shelter
Catalog No. 111009456 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
11-100-9456 10/Pk.
1 options
Catalog No. 11-100-9456 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD196
Add to Cart
Add to Cart
A polypropylene protective outdoor shelter for housing up to four radiello™ samplers during outdoor/ambient air sampling.
The shelter allows for adequate ventilation while simultaneously protecting the samplers from harsh weather conditions. The shelter can be mounted to a variety of fixtures including lamp posts, traffic lights, and telephone poles of various diameters. The shelter can easily be transported from the lab and mounted without the use of tools. Two mounting strips are included with each shelter.
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
For Use With (Application) Air Monitoring
Quantity 10/Pk.
Type Outdoor Shelter
radiello is a trademark of Institi Clinici Scientifici Maugeri