Learn More
Med Vet International Radiographic Foam Positioners, Rectangles and Discs, 4" x 13.5" x 13.5", J1385B, Each
Catalog No. 5000581150
Encompass_Preferred
These high-density polyfoam positioning blocks are comfortable and supportive - Open cell foam in gray/black color - Available in a variety of sizes - Most economical product - Radiolucent Rectangles and Discs 4" x 13.5" x 13.5"
Catalog No. 50-005-81150 Supplier Med Vet International Supplier No. JOR1385B
These high-density polyfoam positioning blocks are comfortable and supportive - Open cell foam in gray/black color - Available in a variety of sizes - Most economical product - Radiolucent Rectangles and Discs 4" x 13.5" x 13.5"