Learn More
Med Vet International Radiographic Vinyl Covered Foam Positioner 6" Disc, J1385CN, Each
Catalog No. 5000581381
Encompass_Preferred
These high-density polyfoam positioning blocks are comfortable and supportive. • Open cell foam covered with blue vinyl • Easy to clean and impervious to fluids • Available in a variety of sizes • Radiolucent   J1385cn 1.5"H x 6" diameter
Catalog No. 50-005-81381 Supplier Med Vet International Supplier No. JOR1385CN
These high-density polyfoam positioning blocks are comfortable and supportive. • Open cell foam covered with blue vinyl • Easy to clean and impervious to fluids • Available in a variety of sizes • Radiolucent   J1385cn 1.5"H x 6" diameter