Learn More
Hach Company Radiometer Analytical CDC749 Conductivity Cell for Microsamples - 2-pole platinized Pt probe, k = 1.7 cm⁻¹, 4 mm lower diameter, glass body, fixed cable with MAB6-style plug; compatible with MeterLab/CDM series
Catalog No. 5000207495 Shop All Hach Company Products
Encompass
The Radiometer Analytical CDC749 is a compact 2-pole conductivity cell designed for precise conductivity measurement of microsamples and small-volume samples in test tubes. It features a platinized platinum electrode, a cell constant of 1.7 cm⁻¹, and a small lower diameter for limited sample volumes. The glass body and robust construction provide chemical compatibility and measurement stability.
  • Designed for microsamples and test-tube measurements.
  • Platinized platinum 2-pole electrode for stable conductivity readings.
  • Cell constant k = 1.7 cm⁻¹ for accurate low-volume measurements.
  • 4 mm lower diameter to fit small sample volumes.
  • Glass body for chemical compatibility and dimensional stability.
  • Fixed cable with a multi-pin plug for direct connection to compatible meters.
  • Supplied with a certificate of conformity and a 12-month warranty.
Catalog No. 50-002-07495 Supplier Hach Company Supplier No. E61M014
The Radiometer Analytical CDC749 is a compact 2-pole conductivity cell designed for precise conductivity measurement of microsamples and small-volume samples in test tubes. It features a platinized platinum electrode, a cell constant of 1.7 cm⁻¹, and a small lower diameter for limited sample volumes. The glass body and robust construction provide chemical compatibility and measurement stability.
  • Designed for microsamples and test-tube measurements.
  • Platinized platinum 2-pole electrode for stable conductivity readings.
  • Cell constant k = 1.7 cm⁻¹ for accurate low-volume measurements.
  • 4 mm lower diameter to fit small sample volumes.
  • Glass body for chemical compatibility and dimensional stability.
  • Fixed cable with a multi-pin plug for direct connection to compatible meters.
  • Supplied with a certificate of conformity and a 12-month warranty.