Learn More
TA Instruments Radiometer Model R2000, Optional Calibration Accessory for PCA, Compatible with Novacure 2100 and Omnicure S2000, Used to Measure and Calibrate PCA Light Intensity
Catalog No. 5000580621 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Radiometer Model R2000 is an optional calibration accessory for the Photocalorimeter Accessory (PCA). It is compatible with both the Novacure 2100 and Omnicure S2000 systems. The radiometer measures and calibrates PCA light intensity to ensure accuracy and reliability.
Catalog No. 50-005-80621 Supplier TA Instruments Supplier No. 200700.001
Waters TA Instruments Radiometer Model R2000 is an optional calibration accessory for the Photocalorimeter Accessory (PCA). It is compatible with both the Novacure 2100 and Omnicure S2000 systems. The radiometer measures and calibrates PCA light intensity to ensure accuracy and reliability.