Learn More
Medchemexpress LLC Radioprotectin-1 100mg | 1622006-09-0 | 100 MG
SDP
Catalog No. 5000227444
Encompass_Preferred
Radioprotectin-1 is a small-molecule, nonlipid agonist selective for the lysophosphatidic acid receptor 2 (LPA2). It is supplied for research use and is used in vitro for receptor activation studies and pharmacology research; the reported EC50 for murine LPA2 is 25 nM.
  • Potent LPA2 agonist with reported EC50 25 nM (murine).
  • Suitable for in vitro receptor activation and signaling studies.
  • High reported purity for reliable experimental results.
  • Available in multiple small research pack sizes for assay work.
Catalog No. 50-002-27444 Supplier Medchemexpress LLC Supplier No. HY114380100MG
Radioprotectin-1 is a small-molecule, nonlipid agonist selective for the lysophosphatidic acid receptor 2 (LPA2). It is supplied for research use and is used in vitro for receptor activation studies and pharmacology research; the reported EC50 for murine LPA2 is 25 nM.
  • Potent LPA2 agonist with reported EC50 25 nM (murine).
  • Suitable for in vitro receptor activation and signaling studies.
  • High reported purity for reliable experimental results.
  • Available in multiple small research pack sizes for assay work.