Learn More
Medchemexpress LLC HY-114380 10mg Medchemexpress, Radioprotectin-1 CAS:1622006-09-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11438010MG
Encompass_Preferred
Medchemexpress, HY-114380 10mg Radioprotectin-1 CAS:1622006-09-0 Radioprotectin-1 is a potent and specific nonlipid agonist of lysophosphatidic acid receptor 2 (LPA2), with an EC50 value of 25 nM for murine LPA2 subtype. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9339