Learn More
Medchemexpress LLC HY-114380 5mg Medchemexpress, Radioprotectin-1 CAS:1622006-09-0 Purity:>98%
SDP
Catalog No. 501932485
Encompass_Preferred
Medchemexpress, HY-114380 5mg Radioprotectin-1 CAS:1622006-09-0 Radioprotectin-1 is a potent and specific nonlipid agonist of lysophosphatidic acid receptor 2 (LPA 2 ) , with an EC 50 value of 25 nM for murine LPA 2 subtype [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2485 Supplier Medchemexpress LLC Supplier No. HY1143805MG
Medchemexpress, HY-114380 5mg Radioprotectin-1 CAS:1622006-09-0 Radioprotectin-1 is a potent and specific nonlipid agonist of lysophosphatidic acid receptor 2 (LPA 2 ) , with an EC 50 value of 25 nM for murine LPA 2 subtype [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.