Learn More
Grainger Radius Gauge Set
Catalog No. 502766600
Encompass_Preferred
The Mitutoyo Radius Gauge Set with Fixed Leaves is a precision measuring tool for checking internal and external radii. It consists of 13 fixed radius leaf gauges ranging from 0.5mm to 3mm in 0.25mm increments. The gauges are made from stainless steel with satin chrome plated finish. They feature a knurled grip area and insertion marks. This radius gauge set is accurate to +/- 0.002mm and conforms to DIN 878 standards. The gauges are housed in a fitted plastic case for storage and portability. This is a high quality radius gauge set well-suited for use in inspection and quality control across many industries including automotive, aerospace, and manufacturing.
Catalog No. 50-276-6600 Supplier Grainger Supplier No. 16X229
The Mitutoyo Radius Gauge Set with Fixed Leaves is a precision measuring tool for checking internal and external radii. It consists of 13 fixed radius leaf gauges ranging from 0.5mm to 3mm in 0.25mm increments. The gauges are made from stainless steel with satin chrome plated finish. They feature a knurled grip area and insertion marks. This radius gauge set is accurate to +/- 0.002mm and conforms to DIN 878 standards. The gauges are housed in a fitted plastic case for storage and portability. This is a high quality radius gauge set well-suited for use in inspection and quality control across many industries including automotive, aerospace, and manufacturing.