Learn More
Grainger Radius Gauge Set
Catalog No. 502767194
Encompass_Preferred
The Mitutoyo Radius Gauge Set with Fixed Leaves is a precision measuring tool for checking internal and external radii. It consists of 16 blades with radius sizes ranging from 0.5mm to 16mm in 0.5mm increments. The blades are made from stainless steel with satin chrome finish and have a thickness of 0.3mm. This radius gauge set allows for quick and accurate measurement and inspection of inner and outer radii on parts and components. The compact case design provides safe storage. This is an ideal inspection tool for use in machining, fabrication, toolmaking, and other manufacturing applications requiring precise radius measurements.
Catalog No. 50-276-7194 Supplier Grainger Supplier No. 16X230
The Mitutoyo Radius Gauge Set with Fixed Leaves is a precision measuring tool for checking internal and external radii. It consists of 16 blades with radius sizes ranging from 0.5mm to 16mm in 0.5mm increments. The blades are made from stainless steel with satin chrome finish and have a thickness of 0.3mm. This radius gauge set allows for quick and accurate measurement and inspection of inner and outer radii on parts and components. The compact case design provides safe storage. This is an ideal inspection tool for use in machining, fabrication, toolmaking, and other manufacturing applications requiring precise radius measurements.