Learn More
RAE Systems Inc Colorimetric Gas Detection Tube, Hydrogen Sulfide, 2.5-60 ppm (Pack of 10)
SDP
Catalog No. NC0910870
Encompass_Preferred
This item has a minimum qty of 11 per supplier requirements.
Easy to read, providing a sharp, clear colour change directly corresponding to the concentration of gas present.No calibration or other maintenance required.Wide assortment of tubes available to detect over 300 gases and vapors.Springless piston design accurately draws 50 and 100 cc volumes.Compatible with LP-1200 piston hand pump, a reliable way to draw samples of an exact volume required for the most accurate measurements.Tight vacuum seal is formed by a greased plunger gasket.Tapered rubber inlet accommodates a range of tube diameters for different types of tubes.Inlet filter prevents glass pieces and dust from entering the shaft.End-of-flow indicator in the handle turns white when the gas sampling is complete.
Catalog No. NC0910870 Supplier RAE Systems Inc Supplier No. 1010310/DEL
This item has a minimum qty of 11 per supplier requirements.
Easy to read, providing a sharp, clear colour change directly corresponding to the concentration of gas present.No calibration or other maintenance required.Wide assortment of tubes available to detect over 300 gases and vapors.Springless piston design accurately draws 50 and 100 cc volumes.Compatible with LP-1200 piston hand pump, a reliable way to draw samples of an exact volume required for the most accurate measurements.Tight vacuum seal is formed by a greased plunger gasket.Tapered rubber inlet accommodates a range of tube diameters for different types of tubes.Inlet filter prevents glass pieces and dust from entering the shaft.End-of-flow indicator in the handle turns white when the gas sampling is complete.