Learn More
RAE Systems Inc RAE 4 GAS CAL GAS 18%O2
SDP
Catalog No. NC9031492
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Four-Gas Calibration Mix ,34L Aluminum Cylinder (50%LEL / 2.5% VOL CH4,18% O2,10 PPM H2S, 50 PPM CO, BAL N2)
Catalog No. NC9031492 Supplier RAE Systems Inc Supplier No. 6000050007
This item has a minimum qty of 2 per supplier requirements.
Four-Gas Calibration Mix ,34L Aluminum Cylinder (50%LEL / 2.5% VOL CH4,18% O2,10 PPM H2S, 50 PPM CO, BAL N2)