Learn More
Aviva Systems Biology Corp RAE1 Antibody - C-terminal region (ARP40301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAE1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RAE1. Alternate names: MIG14, MRNP41, Mnrp41, dJ481F12.3, dJ800J21.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57384