Learn More
Aviva Systems Biology Corp RAET1G Antibody - middle region (ARP65597_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65597_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAET1G. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human RAET1G. Alternate names: ULBP5ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-288-525